Order Entry
Italy
ContactUsLinkComponent
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI
Certificates
undefined
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Frequently Bought Together