Absorbent mats, oil-only, 4 in 1®, PIG®
Supplier: NEW PIG
Certificates
About this item
Highly absorbent mats are made from a eight-layer 100% PP medium-weight construction and function as pads, rolls, socks or wipes. They absorb and retain a variety of leaks, drips, and spills of oils and fuels, except water.
- Scuff-resistant, tear-resistant, even under heavy foot traffic or abrasive conditions
- Flame-resistant - material melts when exposed to high heat
- Dimple pattern wicks of liquid throughout mat for faster and easier clean-up
- Rolls are perforated every 250 mm
Frequently Bought Together