Thermo Scientific Nalgene 3114-0010 Round-bottom tubes; 10 mL; FEP; 2/pack
Catalog # MFLX06327-30
Supplier: Avantor Fluid Handling
Specifications
See all product configurations- Catalog No:
- MFLX06327-30
- MFLX06336-50
- MFLX06328-15
- MFLX06327-42
- MFLX06327-31
- MFLX06338-10
- MFLX06337-11
- MFLX06327-45
- MFLX06327-41
- MFLX06338-50
- MFLX06338-30
- MFLX06324-64
- MFLX06328-10
- MFLX06324-66
- MFLX06328-20
- Autoclavable:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Separate materials without leaks.
- Ideal for high-speed centrifugation qnd lipis extraction
FEP tubes are ideal for high-speed centrifugation of aggressive chemicals. Sealing caps ensure leakproof performance at speeds greater than 10,000 RCF ×g and are recommended when spinning hazardous materials. Tubes are autoclavable adn withstand temperatures from -100 to 150 °C (-148 to 302 °F).
Note: FEP tubes must be filled to 100% of capacity and centrifuges under refrigeration; do not use at room temperature.
Thermo Scientific Nalgene® Nalge Thermo Scientific Nunc International
PTFE, TEFZEL® E.I. DuPont de Nemours & Co.
VITON® Dupont Dow Elastomers L.L.C.
- Cap included:Yes
- Cap material:Tefzel® ETFE
- Tube ext.Ø:16 mm
- Colour:Clear
- Description:Round-bottom tubes; 10 mL; FEP; 2/pack
- Length:81,5 mm
- Material:FEP
- Environmentally Preferable:
- Warranty:1 year
- Autoclavable:Yes
- Cap type:Screw cap
- Graduations:-
- Pk:2
- RCF max.:50000 ×g
- Closure size no.:13
- Tube bottom shape:Round
- Tube capacity:10 ml
Frequently Bought Together