Water for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100
Melting Pt:0
Density:1 g/cm³ (4 °C)
Molecular Weight:18.02 g/mol
Merck Index:13,10098
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Abs.@200nm:0.1
- Abs.@210nm:0.1
- Abs.@254nm:0.005
- Abs.@300nm:0.005
- Ammonia:0.3 ppm
- Calcium Test:Pass
- Carbon Dioxide Test:Pass
- Chloride:0.5 ppm
- DNase:none detected
- Endotoxins:0.25 EU/mL
- Liquid Chromatography Suitability:Pass
- Oxidizable Substances:Pass
- pH @25°C:5.0-7.0
- Protease:none detected
- RNase:none detected
Specification Test Results
Frequently Bought Together