Order Entry
India
ContactUsLinkComponent
VWR®, Tube Rotator
VWR®, Tube Rotator
Supplier:  VWR Collection
Certificates
undefined
VWR®, Tube Rotator
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 444-0502
  • 444-0500
  • Model:
  • Tube rotator
  • Tube rotator
  • Capacity:
  • 36×1,5/2,0 ml tubes or 10×10/15 ml and 16×5/7 ml tubes or 6×50 ml tubes
  • 36×1,5/2,0 ml tubes or 10×10/15 ml and 16×5/7 ml tubes or 6×50 ml tubes
  • Shaking motion:
  • Rotating
  • Rotating
  • Speed:
  • 20 min⁻¹ (fixed)
  • 20 min⁻¹ (fixed)
  • Angle:
  • Plates freely adjustable from horizontal to vertical
  • Plates freely adjustable from horizontal to vertical
  • W×D×H:
  • 239×127×153 mm
  • 239×127×153 mm
  • Operating temperature:
  • 4...65 °C
  • 4...65 °C
  • Weight:
  • 2 kg
  • 2 kg
  • Max. load:
  • 0,8 kg
  • 0,8 kg