Maglumi X3, Fully automatic Chemiluminescence Immunoassay (CLIA) System
SNIB10101003301
: Snibe
- Description:Chemiluminescence Immunoassay (CLIA) System Maglumi X3
- Environmentally Preferable:
- Dimensions:78 (h) x 90 (w) x 75 (d) cm
- Weight:127 kg
- Pk:1
X3 is one of the highest space efficiency CLIA analyzers combines one of the broadest automated CLIA test menus. The comprehensive advanced design of MAGLUMI X3 ensures excellent performance. Intelligent UI interface, user friendly, easy and convenient operation. Intuitive indicator light make users no need to check reagents and consumables frequently.
- Small but powerful, the bthroughput is upto 200 tests/hour, a throughput per unit area is 294T/h/m
- Compatible with all Maglumi Reagents, one of the broadest automated CLIA Test menus in the world (166 test parameters)
- The latest intelligent washing technology and bidirectional temperature control measurement guarantee accurate and reliable results. The comprehensive advanced design of Maglumi X3 ensures excellent performance.
- Single reaction cup can avoid light pollution and increases cuvette utilization, it's integrated packaging can avoid the stuck of the cuvette and destroying the cuvette wall.
- No-pause loading / unloading of reagents/samples/reaction cups without waiting or interrupting tests.
Maglumi X3 is a fully automatic chemiluminescence immunoassay (CLIA) system and powerful with a throughput of 200 tests/hr. The system is compatible with all Maglumi reagents and offers the broadest CLIA test menus (166 parameters). It combines latest intelligent washing technology and bi directional temperature control measurement to guarantee accurate and reliable results.
:
CE certified.