- Description:Hazardous materials cabinet, 90 gal., type: Self-closing doors
- W×D×H:Inside dimension: 1003×769×1536 mm; Outside dimension: 1092×863×1651 mm
- Environmentally Preferable:
- Pk:1
- Weight:206,7 kg
- Storage capacity:340,6 L
Identify and safely store hazardous materials such as acids, bases, oxidizers, or toxic material.
- Ideal for areas with limited floor space or to expand current storage conditions
- Powder-coat paint finish provides increased chemical resistance
- It comes with a ten-year warranty
Deep slimline hazardous material Safety cabinets have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and levelling feet. Self-close doors shut and latches automatically when a fusible link melts at 74 °C (165 °F) under fire conditions. Unique, concealed self-closing mechanism offers obstruction-free access to contents.
Additionally, they include polyethylene trays that sit on top of galvanized steel shelves and a separate polyethylene liner for the bottom sump to resist aggressive chemicals. Easily remove the liner for quick cleaning of drips and leaks. An all epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance.
Each cabinet comes with a 'Hazardous - Keep Fire Away' label, as well as a pack of ten application-specific labels to choose from and apply to them in order to indicate its contents.