Hose Fitting Band Clamps, Stainless Steel
Quick and easy assembly without tools.
- Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.
- Environmentally Preferable:
Frequently Bought Together