Order Entry
India
ContactUsLinkComponent
Betaine enhancer solution 5 M (5X) for PCR
undefined
Betaine enhancer solution 5 M (5X) for PCR
  • Environmentally Preferable:
  • Functional test - 1x Betaine Enchancer Solution is tested for amplification of 613bp fragment from human genomic DNA using Taq DNA polymerase at standard PCR Conditions:
    Passes test
  • Volume check- fill volume >= 0.1ml per tube:
    Passes test
Frequently Bought Together