Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED® (8-12 Mesh), activated, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
CAS Number:70955-01-0
:1318-02-1
MDL Number:MFCD03457537
Density:1.1…2.2 g/cm³ (20 °C)
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 5 %
- Loss on Drying at 150°C:≤5 %
- Bead Size (mm):1.6 - 2.5 mm
Specification Test Results
Frequently Bought Together