Gooch crucibles
Supplier: MORGAN ADVANCED MATERIALS HALD
Certificates
About this item
Porcelain gooch crucibles, with perforated base, glazed.
- For use in temperatures up to 1150 °C
Ideal for total suspended solids determination.
Frequently Bought Together