Order Entry
Ireland
Orders LinkContactUsLinkComponent
Sodium laurate 98%

Sodium laurate 98%

Corrosive
Environment
Sodium laurate 98%
Certificates
Sodium laurate 98%

3 Options Available Below

Product Details & Documents
Synonyms:  Sodium Laurate, Lauric acid sodium salt, Sodium dodecanoate
  • CAS Number:629-25-4
  • Formula:C₁₂H₂₃NaO₂
  • MDL Number:MFCD00041754
  • Molecular Weight:222.304 g/mol
  • Storage Temperature:Ambient
  • EINECS:211-082-4
  • UN:3077
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Water:
    ≤3 %
  • GC:
    ≥97.5 %
  • Sodium (Na):
    7.8 to 15.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 67,20 €
    67,20 €
    1 * 5 g
       67,20 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 208,00 €
    208,00 €
    1 * 25 g
       208,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 658,00 €
    658,00 €
    1 * 100 g
       658,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.