2-Chloro-4-methoxybenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Chloro-p-anisic acid
CAS Number:21971-21-1
:
Formula:C₈H₇ClO₃
MDL Number:MFCD00085943
Molecular Weight:186.59 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together