2,4-Dichloro-5-methoxypyrimidine ≥97%
Catalog # H56153.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
CAS Number:19646-07-2
:
Formula:C₅H₄Cl₂N₂O
MDL Number:MFCD02685670
:179.00 g/mol
:GHS05
:GHS07
Melting Pt:66…70 °C
Storage Temperature:Ambient