Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl
CAS Number:65391-42-6
:
Formula:C₁₆H₂₅ClN₂O₄
MDL Number:MFCD00058004
Molecular Weight:308.4 · 36.5 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together