Acetyl chloride 1 mol/l in dichloromethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Acetyl chloride 1M in Dichloromethane
CAS Number:75-36-5
:
Formula:C₂H₃ClO
MDL Number:MFCD00000719
Boiling Pt:50.9
Molecular Weight:78.5 g/mol
Melting Pt:-112
Flash Pt:5
ADR:6.1 III
Merck Index:13,00087
EINECS:200-865-6
UN:1593
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together