Shelving systems, polymer, MetroMax i™
Supplier: Metro International
New Product
Certificates
About this item
MetroMax i™ shelving offers lifelong durability - the strength of steel and a lifetime warranty against rust and corrosion. Removable polymer shelf mats and smooth surfaces promote easy routine cleaning. Ideal for most environments where stainless steel is desired for its corrosion proof properties.
- Microban® antimicrobial product protection is built into every shelf mat and touch points
- Stationary, mobile, and high density storage options
- Shelving up to 800 lbs. (363 kg) per shelf
Built of advanced polymers with type 304 stainless steel corners.
Frequently Bought Together