Order Entry
Ireland
ContactUsLinkComponent
 
Solubility index tube
Catalog # 614-0170
Supplier:  GERBER FUNKE
undefined
Solubility index tube
Catalog # 614-0170
Supplier:  GERBER FUNKE

Specifications

  • Type:
    for SuperVario N centrifuge
  • Capacity:
    50 ml
  • Description:
    ADPI, graduated from 0-20 ml, mark at 50 ml
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

ADPI, graduated from 0 to 20 ml, mark at 50 ml.