2-Chloro-N,N-dimethylacetamide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2675-89-0
:
Formula:C₄H₈ClNO
MDL Number:MFCD00039360
Boiling Pt:98…100 °C (11 mmHg)
Density:1.182
Molecular Weight:121.57 g/mol
Storage Temperature:Ambient
EINECS:220-224-4
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together