Order Entry
Ireland
ContactUsLinkComponent
Biocide, sterile filtered, Klercide™ Sporicidal Low Residue Peroxide
  141-1202
undefined
Biocide, sterile filtered, Klercide™ Sporicidal Low Residue Peroxide
  141-1202
  • Description:
    Klercide™ Sporicidal Low Residue Peroxide, biocide, sterile filtered
  • Environmentally Preferable:
  • Packed:
    1 litre bottle with trigger spray
  • Pk:
    6
Suitable for ISO Class 5, Grade A and B (microbial count) areas: cleanroom processed and sterile, double bagged. Blend of 6% hydrogen peroxide and deionised water, with broad spectrum activity including bacterial spores. Filtered to 0,2 µm.
  • CEN and isolate tested
  • Safe and effective; non toxic and non corrosive
  • Fully compatible and ideal for a rotational regime
  • SteriShield Delivery System™ ensures 3 months in-use shelf life
 : EN standards: EN 1276 - bactericidal activity, EN 1650 - fungicidal activity, EN 13697 - bactericidal surface test, EN 13704 - sporocidal activity, EN 14476 - virucidal.
 : Use biocides safely. Always read the label and product information before use.