Potassium nitrate E252, FCC, BP, Ph. Eur., USP, extra pure, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Nitrate of Potash
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
Density:2.109 g/cm³ (20 °C)
Molecular Weight:101.1 g/mol
Melting Pt:334
ADR:5.1 III
Merck Index:13,07733
EINECS:231-818-8
UN:1486
REACH:01-2119488224-35
:GHS03
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together