4-Methylcyclohexanone for synthesis, Sigma-Aldrich®
: Merck
:589-92-4
:
:C₇H₁₂O
:MFCD00001643
:170
:–41 °C
:0.92 g/cm³ (20 °C)
:112.17 g/mol
:48
:2297
:3 III
:M-Clarity™ quality level = MQ200
:209-665-3
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) practically insoluble
- Melting Point:-41 °C
- Molar Mass:112.17 g/mol
- Boiling Point:169 - 171 °C (1013 hPa)
- Vapor Pressure:20 hPa (65 °C)
- Flash Point:48 °C
- Density:0.92 g/cm3 (20 °C)
Frequently Bought Together