Benzoin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Hydroxy-2-phenylacetophenone
CAS Number:119-53-9
:
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00004496
Molecular Weight:212.25 g/mol
:M-Clarity™ quality level = MQ200
EINECS:204-331-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.3 g/l (25 °C)
- Melting Point:132 - 134 °C
- Molar Mass:212.24 g/mol
- Bulk Density:350 kg/m3
- Boiling Point:344 °C (1023 hPa)
- Vapor Pressure:1.3 hPa (136 °C)
Specification Test Results
Frequently Bought Together