Order Entry
Ireland
ContactUsLinkComponent
Microgen® Listeria ID
Microgen® Listeria ID
Catalog # MIBIMID-67
CAS Number:  
undefined
Microgen® Listeria ID
Catalog # MIBIMID-67
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Microgen® Listeria ID kit
  • Environmentally Preferable:
Specifications
About this item
Developed specially for the food industry and conforming to the requirements of all international food standards, this identification contains a unique micro-haemolysin test providing a fully self-contained identification system.
  • AFNOR validated
  • No sub-culture necessary from selective media
  • AOAC-RI validated
  • CMMAS assessed and approved
For the identification of all Listeria spp. (six species). The above organisms can be identified from selective agar or non-selective agar.