Blood collection needles, BD Vacutainer®
Supplier: BD Medical
Certificates
About this item
BD Vacutainer® blood collection needles have an advanced low-angle bevel design and are coated with silicone, a low friction lubricant, ensuring very gentle and smooth vein entry.
- Individual laser quality control maintains the highest quality levels
- Suitable for use with multiple tube samples
BD Vacutainer® Luer adapters are sterile devices with a multi-sampling valve and are designed for use with a catheter or hypodermic needle to collect blood with blood collection tubes. They are also ideal for use with any female Luer fitting.
Frequently Bought Together