Accessories for Vacuum filtration apparatus, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Vacuum and pressure pump: For use in the fields of microbiological quality control, analyses, medicine and production technology. The pump is used for pumping gases, taking samples (even liquids in a vacuum) and evacuating vessels.
- AC model
- Contamination-free pumping of air, gases and vapours
- Extremely quiet and smooth running
- Equipped with thermo switch and standard fuse
- Oil-free membrane pump
- Delivery m³/h: 3,6 L/min, vacuum: <100 mbar absolute, pressure: 4 bar, weight: 11 kg
Witt's bottle: For filtrate collection in an inserted container. Borosilicate glass bottle, 100×160 mm, with replaceable round lid and side-mounted tubing nozzle for vacuum tubing 8 mm (inside diameter).
Frequently Bought Together