Anthracene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-12-7
:
Formula:C₁₄H₁₀
MDL Number:MFCD00001240
Boiling Pt:340
Melting Pt:111…112 °C
Density:1.24 g/cm³ (20 °C)
Molecular Weight:178.23 g/mol
Flash Pt:121 °C (closed cup)
Merck Index:13,00687
:M-Clarity™ quality level = MQ100
EINECS:204-371-1
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together