Reservoir bottles for automatic pipette / tilt measures
Catalog # BRND126963
Supplier: Brand
Specifications
- NS:29/32
- Capacity:1000 ml
- Description:Reservoir bottle for bottle top tilt measures
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Soda-lime glass.
Capacity 1000 ml
Neck size 29/32
Ordering information: Please order dispenser/tilt measure separately.