0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
n-Nonane
n-Nonane ≥99%
n-Nonane ≥99%
:
Thermo Fisher Scientific
n-Nonane ≥99%
:
Thermo Fisher Scientific
:
111-84-2
:
:
H₃C(CH₂)₇CH₃
:
MFCD00009574
:
150…152 °C
:
–53 °C
:
0.718
:
128.26 g/mol
:
31 °C (87 °F)
:
Ambient
:
203-913-4
:
1920
:
3 III
:
GHS08
:
GHS02
:
GHS07
Environmentally Preferable:
msds_ALFAA16177
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
E/E 96% 96 v/v, TechniSolv®, pure
: VWR Chemicals
Sodium chloride, EMSURE® Reag. Ph. Eur., ACS, ISO analytical reagent, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Reag. USP, ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Tetrahydrofuran stabilised, EMSURE® Reag. Ph. Eur., ACS analytical reagent, Supelco®
: Merck
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
: VWR Collection