Caprylic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid
CAS Number:124-07-2
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00004429
Boiling Pt:238 °C (760 mmHg)
Melting Pt:16…16.5 °C
Density:0.91 g/cm³
Molecular Weight:144.21 g/mol
Flash Pt:110 °C
UN:3265
ADR:8 III
Merck Index:13,01771
EINECS:204-677-5
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4300 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together