Octyl acetate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Acetic acid octyl ester
CAS Number:112-14-1
:
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00009562
Boiling Pt:211 °C (760 mmHg)
Melting Pt:-38.5
Density:0.868 g/cm³
Molecular Weight:172.27 g/mol
Flash Pt:86 °C
Storage Temperature:Ambient
EINECS:203-939-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4175 to 1.4195 (20°C, 589 nm)
- Water:≤0.3 % (K.F.)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together