2-Methylthiobenzamide
Supplier: FLUOROCHEM
Certificates
About this item
Synonyms: 2-Methylbenzothioamide, Thio-o-toluamide
CAS Number:53515-19-8
:
Formula:C₈H₉NS
MDL Number:MFCD03945350
Molecular Weight:151.23
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together