1-Bromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1-Hexyl bromide
CAS Number:111-25-1
:
Formula:BrH₂C(CH₂)₄CH₃
MDL Number:MFCD00000271
Boiling Pt:155
Melting Pt:–85 °C
Density:1.167 g/cm³ (20 °C)
Molecular Weight:165.07 g/mol
Flash Pt:47
Storage Temperature:Ambient
UN:1993
ADR:3 II
:M-Clarity™ quality level = MQ200
EINECS:203-850-2
:GHS02
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:- 1 g/l (20 °C)
- Melting Point:-85 °C
- Molar Mass:165.07 g/mol
- Boiling Point:154 - 158 °C
- Vapor Pressure:4 hPa (20 °C)
- Flash Point:47 °C
- Refractive Index:1.4478 (20 °C, 589 nm)
- Density:1.18 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together