Order Entry
Ireland
ContactUsLinkComponent
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Silica gel, granulated 1 - 3 mm drying agent, with moisture indicator (orange gel), Supelco®
 :  Merck
Silica gel, granulated 1 - 3 mm drying agent, with moisture indicator (orange gel), Supelco®
 :  Merck
  • Environmentally Preferable:
  • Water absorption capacity (24 hrs.,80 % relative humidity):
    ≥ 25.0 %
  • Loss on drying (140 °C):
    ≤ 2.0 %
  • Bulk density:
    about 70
Frequently Bought Together