Hydrogen peroxide 30%, (Perhydrol®) EMSURE® ISO for analysis, Supelco®
: Merck
:7722-84-1
:
:H₂O₂
:MFCD00011333
:∼107 °C (1013 hPa)
:–25.7 °C
:1.11 g/cm³ (20 °C)
:34.01 g/mol
:231-765-0
:2014
:5.1 (8) II
:14,04798
:M-Clarity™ quality level = MQ300
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
- Assay (Manganometric):30% min.
- Color:10 Hazen max.
- Other specifications on request:107 °C (1013 hPa)
- Vapor Pressure:18 hPa (20 °C)
- Density:1.11 g/cm3 (20 °C)
- pH:2 - 4 (H2O, 20 °C)
- Grade:ACS,ISO
- Synonyms:Perhydrol
Frequently Bought Together