2,6-Dichlorophenolindophenol sodium salt hydrate ACS, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: DPIP, 2,6-Dichloroindophenol sodium hydrate, DCIP, Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide hydrate, Sodium 2,6-dichloroindophenolate hydrate, Tillmans' reagent hydrate, 2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt hydrate, 2,6-Dichloroindophenol, sodium salt hydrate, 2,6-Dichlorophenolindophenol sodium salt xhydrate
CAS Number:1266615-56-8
:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂·xH₂O
MDL Number:MFCD00150014
Molecular Weight:290.08 g/mol (anhy)
Melting Pt:> 300 °C
:M-Clarity™ quality level = MQ200
EINECS:210-640-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together