Order Entry
Ireland
ContactUsLinkComponent
Secchi disc
Secchi disc
Catalog # 707-0005
Supplier:  Hach
undefined
Secchi disc
Catalog # 707-0005
Supplier:  Hach
Specifications
  • Description:
    Secchi disc
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The disc is designed to measure water transparency in situ via the semi-quantitative method.
  • Disc slowly lowered in the water until the pattern is no longer visible
  • Depth of immersion is then evaluated
  • Measure related to the transparency
For use in oceans and lakes.
Ø = 200 mm
Certifications: ISO 7027