Order Entry
Ireland
ContactUsLinkComponent
Pharmacy refrigerators, Lec
undefined
Pharmacy refrigerators, Lec
  • Catalog No:
  • 471-0568
  • 471-0817
  • PE102
  • 471-0583
  • 471-0818
  • 471-0569
  • GLNDPSRC273
  • 471-0582
  • 471-0586
  • Model:
  • PG 102C
  • PE 307C
  • PE 102C
  • PE 207C
  • PG 307C
  • PG 1607C
  • PSRC R600a
  • PE 1607C
  • PG 207C
  • Cat. No.:
  • 471-0568
  • 471-0817
  • 472-1045
  • 471-0583
  • 471-0818
  • 471-0569
  • GLNDPSRC273
  • 471-0582
  • 471-0586
  • Air circulation:
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Display:
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Audible and visual temperature alarm
  • Shelves/drawers:
  • 2 wire shelves
  • 2 wire shelves, 1 wire drawer
  • 2 wire shelves
  • 2 wire shelves, 1 wire drawer
  • 2 wire shelves, 1 wire drawer
  • 5 wire shelves
  • 5 wire shelves
  • 5 wire shelves
  • 2 wire shelves, 1 wire drawer
  • Door type:
  • 1 glass swing door
  • 1 solid swing door, reversible hinge
  • 1 solid swing door, reversible hinge
  • 1 solid swing door, reversible hinge
  • 1 glass swing door
  • 1 glass swing door
  • 1 solid swing door
  • 1 solid swing door, reversible hinge
  • 1 glass swing door
  • Door lock:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Weight:
  • 25,5 kg
  • 32 kg
  • 23,5 kg
  • 28 kg
  • 37 kg
  • 106 kg
  • 60 kg
  • 96 kg
  • 30 kg
Frequently Bought Together