Handheld UV lamps
Supplier: UVP ULTRA VIOLET PRODUCTS
Certificates
About this item
These lightweight lamps can be used with the optional lamp stand for hands-free operation. Replacement tubes are also available.
- Lightweight for easy handling
- Durable plastic housing
- Six watts
- Models available with short wave, long wave UV or a combination short wave/long wave version
- Combination short wave/long wave version comes with a snap-on wavelength selector for switching between wavelengths (UVGL-58 only)
W×D×H: 81×378×64 mm
Weight: 0,9 kg
Frequently Bought Together