Order Entry
Ireland
ContactUsLinkComponent
 
Striped floor marking cleanroom tapes
  819-1700
 :  ULTRATAPE
undefined
Striped floor marking cleanroom tapes
  819-1700
 :  ULTRATAPE

 

  • Width:
    50,8 mm
  • Colour:
    Black/yellow
  • Description:
    Cleanroom tape, floor marking
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    3 Roll

 

 

These striped floor marking cleanroom tapes are made of polyester overlaminated on vinyl with rubber adhesive.

  • Designed for cleanroom use
  • Chemical resistant, withstands heavy traffic and regular wiping and mopping
  • Long-term durability, colours do not fade
  • Individually inspected
  • Packaged and sealed in certified MDPE bags to prevent contamination

They can be used for floor marking and colour coding. Cleanroom coloured and striped hazard warning tapes help designate the OSHA critical areas in cleanrooms.

Length: 16,4 m

 : Supplied double bagged.