Order Entry
Ireland
ContactUsLinkComponent
Adhesive plate sealing film for qPCR, ABsolute™
Adhesive plate sealing film for qPCR, ABsolute™
  732-0498
undefined
Adhesive plate sealing film for qPCR, ABsolute™
  732-0498
  • Optical property:
    Optically clear
  • Description:
    ABsolute™ qPCR sealing film, sheets
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    50
Optically clear adhesive plate sealing film designed for qPCR and other fluorescence applications.
  • Strong pressure sensitive adhesive on film prevents evaporation during thermal cycling or sample storage
  • Peelable with no sticky residue left on the plate
  • End tabs for easy seal application and removal
  • Seal recommended for use in conjunction with a cycler that has a screw-down or clip-down lid