PCR plates, 96- and 384-well
PP, non-sterile, without lid. The use of the 96-well format allows the scale up of basic PCR work, while the 384-well format is ideal for high throughput screening projects, such as the sequencing of complex genomes.
- Ultra thin PP for optimal heat transfer
- Available in different designs, including half-skirted 96-well plates and full-skirted 384-well plates with one notch, suitable for ABI systems
- Alphanumeric coding on 384-well plates, printed in colour
- Free of DNase, RNase and human DNA, non-pyrogenic
Colour: Natural
Colour: Natural
- Catalog No:
- 731-0040
- 731-0037
- 731-3016
- 731-0039
- GREI652260_100
- GREI652270_100
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Frequently Bought Together