Order Entry
Ireland
ContactUsLinkComponent
MPLC cartridge columns, Brownlee™
MPLC cartridge columns, Brownlee™
Supplier:  PerkinElmer
Certificates
MPLC cartridge columns, Brownlee™
Supplier:  PerkinElmer
Specifications
  • Catalog No:
  • PERC07110019
  • PERC07110020
  • PERC07110033
  • PERC07110037
  • PERC07110016
  • Carbon load:
  • 14%
  • 14%
  • N/A
  • 3%
  • 11%
  • Endcapped:
  • Yes
  • Yes
  • N/A
  • No
  • Yes
  • Surface area:
  • 180 m²/g
  • 180 m²/g
  • 180 m²/g
  • 180 m²/g
  • 180 m²/g
Specifications
Guard cartridge holders for Brownlee™ cartridges
Product Information