Laboratory coats, flame retardant, cotton/Proban
These coats are made from a Proban treated cotton drill fabric.
- Material does not ignite when in direct contact with flame, but forms a blackened char
Colour: white
: Other sizes are available, please contact your local Avantor sales office for further information.
- Catalog No:
- 113-2202
- 113-2205
- 113-2203
- 113-2204
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- Cotton
- Cotton
- Cotton
- Cotton
Frequently Bought Together