Jerricans, Narrow Neck, HDPE
: RIXIUS
New Product
HDPE, translucent.
- Good chemical resistance
- Temperature resistant from −40 to +110 °C
- UN canister with Y approval
Universal canister for industry, trade, and laboratories. Ideal for packaging, storage and transportation. With UN or Y approval.
: Canisters are supplied with or without screw cap. Additional compatible screw cap types are available separately.
Screw caps
Frequently Bought Together