Jars, low form, with Twist-off thread
Supplier: RIXIUS
Certificates
About this item
Glass, without cap.
- Good chemical and physical properties
- For use in applications without high thermal loads
- Fitting metal screw caps are available under codes 216-3630 to 216-3633
Perfect for sample collection, storing and transporting of powders, granulates, pastes or liquids.
Caps
Product Information
Frequently Bought Together