Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Chromogranin B Mouse Monoclonal Antibody [clone: CHGB/4560]
  ANTIA316170-100
New Product
Anti-Chromogranin B Mouse Monoclonal Antibody [clone: CHGB/4560]
  ANTIA316170-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CCB peptide
  • Antigen symbol:
    Chromogranin B
  • Clonality:
    Monoclonal
  • Clone:
    CHGB/4560
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05060
  • Antigen synonyms:
    Cg B|Chromogranin B|SCG1_HUMAN|SCG1|CgB|ChromograninB|Chgb|Secretogranin 1|Chromogranin-B
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    76 kDa/110 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 250-450) of human CHGB protein. The exact sequence is proprietary.
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [CHGB/4560] antibody to Chromogranin B for ELISA with samples derived from human.

  • Validated applications: ELISA

Type: Primary
Antigen: Chromogranin B
Clonality: Monoclonal
Clone: CHGB/4560
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human