Order Entry
Ireland
ContactUsLinkComponent
Human GDX ELISA Kit
Human GDX ELISA Kit
  ANTIA302814-96
New Product
 :  
undefined
Human GDX ELISA Kit
  ANTIA302814-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    GDX
  • Description:
    Human GDX ELISA kit
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Human GDX ELISA kit exhibits high specificity and excellent specificity for the detection of human GDX. No significant cross-reactivity or interference between GDX and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,094 ng/ml
  • Regulatory status:
    RUO

 

 

Human GDX ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GDX in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human GDX ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human GDX in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for GDX has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GDX present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-GDX antibody, which binds the captured GDX present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of GDX captured in each well. The concentration of GDX can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.