Optical Glass Filters
Supplier: SCHOTT AG Lighting and imaging
Certificates
About this item
Optical glass filters are used to change the light color of the emitted light from a KL Cold lightsource.
- Insert Filters for cold light source with filter holder
- Glass Optical Filters for better temperature resistance
- Color filters with high transmission and unaffected by light entrance angle
- The Daylight Filter for Halogen Light Sources Increases the color temperature from a warm halogen light to a cold daylight
- The Halogen Filter for LED lightsources decrease the color temperature from a cold daylight to a warm halogen light
- The (empty) Filter Holder for insert Filters can be used when using multiple filters for easier installation
Optical Color Filters are used to check color printed samples, for instance, to assess the print quality of packaging. When using colored samples, color filters allow you to reduce colors that match the color of the filter. Conversely, other colors can be emphasized. Place Insert Filters inside the cold light source to evenly illuminate the entire fiber bundle. These will then work with all types of fiber optics. Can be used with all standard fiber optics for the respective light source.
Ordering information: Also available are screw-on filters for the focus lenses of goosenecks and flexible light guides.
Insert filters
Product Information
Frequently Bought Together