Anti-PPP3CB Rabbit Polyclonal Antibody
Catalog # PRSI19-109
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Protein phosphatase 3 catalytic subunit beta
- Antigen symbol:PPP3CB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5532
- Antigen synonyms:beta isoform|CALNA2|protein phosphatase 3 (formerly 2B)|serine/threonine-protein phosphatase 2B catalytic subunit beta isoform|CAM-PRP catalytic subunit|CNA2|protein phosphatase from PCR fragment H32|catalytic subunit|calcineurin A beta|calcineurin A2|CNA beta|beta isozyme|calmodulin-dependent calcineurin A subunit beta isoform|protein phosphatase 2B|CALNB|PP2Bbeta|protein phosphatase 3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 286-525 of human PPP3CB (NP_001135825.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PPP3CB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse